Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CARD10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. p-7184249
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111422 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111422 Supplier Invitrogen™ Supplier No. PA5111422
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LTTLTSLEGT KALLEVQLQR AQGGTCLKAC ASSHSLCSNL SSTWSLSEFP SPLGGPEATG EAAVMGGPEP HNSEEATDSE KEINRLSILP F Highest antigen sequence indentity to the following orthologs: Mouse - 89%, Rat - 87%.

CARMA proteins belong to the membrane-associated guanylate kinase-like (MAGUK) family of proteins that can function as molecular scaffolds that assist assembly of signal transduction molecules. CARMA1, CARMA2, and CARMA3 share high degrees of sequence and functional homology, but their tissue-specific distribution suggests that they serve distinct biological functions in different cell types. As with CARMA1, the CARD domain of CARMA3 has been shown to specifically interact with BCL10, a protein known to function as a positive regulator of cell apoptosis and NF-Kappa-B activation. When expressed in cells, this protein binds to BCL10 and activates NF-Kappa-B Recent experiments have shown that CARMA3 is required for EGF-induced NF-Kappa-B activation and contributes to tumor growth in vivo, suggesting that CARMA3 may serve as a new therapeutic target for the treatment of EGFR-driven tumors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CARD10
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CARD10
Gene Accession No. Q9BWT7
Gene Alias AI449026; Bcl10 binding protein and activator of NFKB; bcl10-interacting MAGUK protein 1; Bimp1; Card10; CARD-containing MAGUK 3 protein; CARD-containing MAGUK protein 3; Carma 3; Carma3; caspase recruitment domain family member 10; caspase recruitment domain family, member 10; caspase recruitment domain-containing protein 10
Gene Symbols CARD10
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human CARD10. Recombinant protein control fragment (Product #RP-106649).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29775
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.