Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heathrow Scientific™ Cardboard Tube Storage Box and Lid
GREENER_CHOICE

Catalog No. 17100425
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-100-425 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-100-425 Supplier Heathrow Scientific™ Supplier No. HS2860M
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Cardboard Tube Storage Box offers a quick and inexpensive way to organize your tubes with the added convenience of disposing after use.

  • Write-on surface for descriptions or self-numbering systems
  • Low Temp: -196°C

Specifications

Color White
Material Cardboard
Shape Square
Type Freezer Box
Holds 15 and 50 mL Tubes (no insert)
Withstand Temperature (Metric) -196°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.