Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cardiac Troponin I Monoclonal Antibody (C5)
GREENER_CHOICE

Catalog No. PIMA120118
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA120118 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA120118 Supplier Invitrogen™ Supplier No. MA120118
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-20118 detects cardiac troponin 1 in human samples.

Cardiac Troponin I (Cardiac-specific troponin I, TnI, Troponin I) is an integral inhibitory protein in cardiac muscle that exists as part of a complex with troponin C (TnC) and troponin T (TnT). The TnT subunit of troponin binds to tropomyosin-t form, a troponin-tropomyosin complex, anchored in place by the binding of TnI to actin, within muscle thin filaments. Structural change resulting from the binding of calcium to specific sites on the regulatory TnC subunit, releases the inhibitory region of TnI from actin, anabling the attachment of the molecular motor protein myosin, allowing for muscle contraction and hence movement. The measurements of blood TnI and TnT levels is an important diagnostics indicator of heart muscle damage, and can be used to differentiate between angina and myocardial infarction in patients with chest pains. Cardiac Troponin I has also been shown to inhibit angiogenesis in vivo and in vitro. Mutations in the Cardiac Troponin I gene cause familial hypertrophic cardiomyopathy type 7 (CMH7) and familial restrictive cardiomyopathy (RCM).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cardiac Troponin I
Applications Affinity Purification, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone C5
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene TNNI3
Gene Accession No. P08057, P19429, P23693, P48787, Q863B6, Q8MKD5
Gene Alias alternatively spliced; cardiac ankyrin repeat kinase; Cardiac muscle troponin T; cardiac troponin I; cardiac troponin I isoform; cardiac troponin I-interacting kinase; cardiac troponin T; cardiac troponin T isoform 1; cardiac troponin T isoform 2; cardiac troponin T2; cardiomyopathy, dilated 1D (autosomal dominant); cardiomyopathy, dilated 2A (autosomal recessive); cardiomyopathy, hypertrophic 2; CARK; CCDD; CMD1D; CMD1FF; CMD2A; CMH2; CMH7; CMPD2; cTnI; cTnT; Ctt; CTTG; D830019J24Rik; LVNC6; MGC116817; RATCTTG; RCM1; RCM3; Serine/threonine-protein kinase TNNI3K; Tn1; TnI; TNNC1; Tnni3; TNNI3 interacting kinase; TNNI3-interacting kinase; TNNI3K; tnni3k {ECO:0000250; TNNT2; Tnnt3; TnTc; Tpn1; troponin 1, type 3; troponin I type 3 (cardiac); troponin I, cardiac 3; troponin I, cardiac muscle; troponin I, cardiac muscle-like; troponin I3, cardiac type; Troponin T cardiac; troponin T cardiac isoform; troponin T isoform 1; troponin T isoform 2; troponin T splicing mutant isoform 1; troponin T splicing mutant isoform 2; troponin T type 2 (cardiac); troponin T, cardiac muscle; troponin T2, cardiac; troponin T2, cardiac type; truncated cardiac troponin T; UniProtKB:Q59H18}
Gene Symbols TNNI3
Host Species Mouse
Immunogen Free human cardiac troponin I, native human cardiac troponin complex, or human cardiac troponin I peptides.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100049696, 100860775, 21954, 29248, 403566, 493744, 511094, 7137
Target Species Human, Mouse, Rat, Canine, Rabbit, Bovine, Pig, Feline, Fish, Goat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.