Learn More
Cardinal Health™ Standard Lab Coats
Catalog No. 191304886 Shop All Cardinal Health Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
Color:
Blue
White
Collar Type:
Knit
Traditional
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Color Collar Type
19-130-4886 2X-Large White Traditional
19-130-1863B Medium Blue Traditional
19-130-1863C Large Blue Traditional
19-130-5288 Medium White Knit
19-130-5289 Large White Knit
19-130-5290 X-Large White Knit
19-130-5291 2X-Large White Knit
19-130-1863D X-Large Blue Traditional
19-130-1863E 2X-Large Blue Traditional
19-130-5281 Medium White Traditional
19-130-5285 X-Large White Traditional
19-130-5283 Large White Traditional
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Catalog No. 19-130-4886 Supplier Cardinal Health™ Supplier No. 2304LC
This item is not returnable. View return policy
Standard lab coats protect from low fluid levels. Choose from blue or white with traditional or knit collar.
Made of high-quality, fluid-resistant, tri-layer spunbond meltblown spunbond (SMS) material. White or blue.
  • Traditional or knit collar
  • Three pockets
  • Snap-front closure
  • Color: white or blue
For industrial use only.
Specifications
Product Type General Purpose Lab Coat
Size 2X-Large
Gender Unisex
Material SMS
Color White
Closure Type Snaps
Cuff Style Knit
Disposable Limited-use
Sterility Non-sterile
No. of Pockets 3
Collar Type Traditional
Model 2304LC
Length (English) 45 in.
Length (Metric) 114.3 cm