Learn More
Description
- Feature full-length front zipper and elastic wrists and ankles
- Basic style offers set-in sleeves and traditional collar
- Premier style offers raglan sleeves, bound collar, and a more generous cut
- Both styles are white
Order Info
To ensure cleanliness, coveralls are packed in double poly-lined bags.
Specifications
Specifications
| Product Type | Cleanroom Coveralls |
| Size | 3X-Large |
| Material | Tyvek |
| Color | White |
| Disposable | Single-use |
| Sterility | Non-sterile |
| For Use With (Application) | Controlled Environments |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.