Description
For use with low-temperature sterilization and applications requiring EO.
- Breathable Tyvek™ material facilitates faster aeration and easier sterilization
- Resistant to puncture and tearing
Specifications
Specifications
| Closure Type | Self-seal |
| For Use With (Application) | Excellent choice for devices requiring low temperature sterilization, produced without an indicator for all types of low temperature sterilization, including EO and plasma |
| Material | Tyvek |
| Dimensions (L x W) | 7.6 x 17.9cm (3 x 7 in.) |