Description
For use with low-temperature sterilization and applications requiring EO.
- Breathable Tyvek™ material facilitates faster aeration and easier sterilization
- Resistant to puncture and tearing
Specifications
Specifications
| Closure Type | Self-seal |
| Material | Tyvek |
| Dimensions (L x W) | 15.2 x 25.4cm (6 x 10 in.) |