Learn More
Echelon Biosciences Research Labs CARDIOLIPIN 16:0 100ug
SDP
Supplier:  Echelon Biosciences Research Labs LC160100UG
Encompass
Cardiolipin (CL) is an important component in prokaryotic and in eukaryotic inner mitochondrial membrane. In humans, CL is made by cardiolipin synthase from phosphatidylglycerol and cytidinediphosphatediacylglycerol. Due to its four acyl chains and small head group, CL can organize into domains which can be proton sinks, fill cavities at protein interfaces, stabilize protein oligomers, and participate in high-curvature membrane regions. Autoantibodies to CL have been found in patients with anti-phospholipid syndrome and lupus. This item is non returnable
Catalog No. 50-822-33