Learn More
Description
Product Features:
- Auscultation is an essential clinical skill needed to assess and monitor patients' conditions
- The SAM II™ trainer can be used with any stethoscope
- All sounds and videos are recorded from live patients for students to experience a life-like simulation
- Users can also create and save their own case videos
- When connected to the included laptop with pre-installed software, a variety of sounds, videos and lessons recorded can be accessed
- When connected to speakers (Sold Separately) SAM II Student Auscultation Manikin can be easily moved into a classroom or an auditorium for group instruction
- The programmable and password protected lectures allow teaching institutions to have numerous instructors utilize the SAM II trainer
Delivery content:
- Portable SAM II Male Auscultation Torso
- New DELL laptop computer pre-installed with auscultation teaching software, programmable and password protected lectures
- Digital copy of SAM`s Lesson Guide: located on the laptop in digital form, can be printed and used to support auscultation teaching anytime
- One SAM t-shirt
- User's manual
How is SAM II Student Auscultation Manikin different from other auscultation products available on the market?
- It is lightweight & portable
- It has the largest library of sounds (24 sounds) & videos (36 videos altogether)
- It has the complete Lesson Guide
- It can be used with any stethoscope
- It has a palpable carotid pulse
- Users can setup password protected lectures for student assessment
- It is available in light and dark skin version
Specifications
Specifications
| Product Type | Manikin |
| For Use With (Application) | Teaching and Learning Auscultation |
| Color | Dark Skin |
| Includes | SAM II Male Auscultation Torso, Dell Laptop with Pre-Installed Auscultation Teaching Software, SAM's Lesson Guide, SAM T-Shirt, User's Manual |
| Weight (English) | 30.20 lb. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

