Learn More
AdipoGen Carfilzomib [PR-171] (1 mg)
SDP
Supplier:  AdipoGen AGCR13669M001
Encompass_Preferred
Carfilzomib [PR-171] (1 mg), AdipoGen Life Sciences. Chemical. CAS: 868540-17-4. Formula: C40H57N5O7. MW: 719.9. Synthetic. Potent and irreversible second-generation peptide epoxyketone class proteasome inhibitor. Synthetic analog of the microbial product epoxomcin. Targets the chymotrypsin-like beta5 subunit of the constitutive 20S proteasome (IC50=5.2nM) and the beta5i subunit [LMP7] of the 20S immunoproteasome (IC50=14nM), with minimal crossreactivity to other proteases. Displays equal potency but greater selectivity for the chymotrypsin-like activity of the proteasome compared to bortezomib. Anticancer compound effective against multiple myeloma in vivo. In vitro, induces cell cycle arrest and apoptosis in human cancer cell lines including multiple myeloma, lymphoma, and various solid tumors (IC50s=2.4-20nM).
Catalog No. 50-149-0157
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.