Learn More
Bioworld Cargille Type B Immersion Oil, 100 mL
Supplier:  Bioworld 217500021
Encompass_Preferred
Item# 739202Cargille has been serving microscopists since 1940 when it introduced the first non-drying immersion oil formula. Since then, they have continued to improve and expand their line of immersion oils to become the undisputed leaders in immersion oil technology. Oils are available in a number of different formulations and can be readily mixed to create intermediate viscosities as required. Types B offers a viscosity of 1250 cSt, and is used in normal light microscopy. The viscosity B type is used as the gap between the cover glass and objective or condenser and slide increases.
Catalog No. 50-199-061
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.