Learn More
TARGETMOL CHEMICALS INC Carglumic Acid 1G
SDP
Supplier:  TARGETMOL CHEMICALS INC T07551G
Encompass_Preferred
Also available in 1 mL, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Carglumic Acid (Ureidoglutaric acid) is an orphan drug used to treat hyperammonemia in patients with N-acetyl glutamate synthase absence. This rare hereditary disease results in elevated blood levels of ammonia, which can eventually cross the blood-brain barrier and lead to neurologic problems, coma, cerebral edema, and death. Carglumic acid was approved by the U.S. FDA on 18 March 2010. Purity 99.8%
Catalog No. 50-288-0666
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.