Learn More
Pyramex™ Carhartt™ Rockwood™ Safety Eyewear
Catalog No. 19155268 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Description:
Capture Clam
Polybag
Lens Tint:
Bronze
Clear
Gray
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Lens Tint
19-155-268 Capture Clam Bronze
19-155-269 Polybag Gray
19-155-270 Capture Clam Gray
19-155-266 Polybag Clear
19-155-267 Polybag Bronze
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-155-268 Supplier Pyramex™ Supplier No. CHB718DTCC
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Anti-fog lenses
  • Suspended anti-fog lens
  • Adjustable rubber nosepiece and co-injected temple tips
  • Available in a Carhartt capture clam or polybag
  • Specifications
    Certifications/Compliance ANSI Z87.1
    Description Capture Clam
    Lens Tint Bronze
    Frame Color Black, Tan
    Weight (Metric) 24.6 g
    Lens Material Polycarbonate
    Frame Material Polycarbonate