Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CARKD Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552705
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552705 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552705 Supplier Invitrogen™ Supplier No. PA552705
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PNAVHEVEKW LPRLHALVVG PGLGRDDALL RNVQGILEVS KARDIPVVID ADGLWLVAQQ PALIHGYRKA VLTPNHVEFS RLYDAVLRGP MDSDDSHGSV LRLSQALGNV TVVQ Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 80%.

Catalyzes the dehydration of the S-form of NAD(P)HX at the expense of ATP, which is converted to ADP. Together with NAD(P)HX epimerase, which catalyzes the epimerization of the S- and R-forms, the enzyme allows the repair of both epimers of NAD(P)HX, a damaged form of NAD(P)H that is a result of enzymatic or heat-dependent hydration. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CARKD
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NAXD
Gene Accession No. Q8IW45
Gene Alias 0710008K08Rik; 2810407E01Rik; ATP-dependent (S)-NAD(P)H-hydrate dehydratase; ATP-dependent (S)-NAD(P)H-hydrate dehydratase {ECO:0000255; ATP-dependent NAD(P)H-hydrate dehydratase; ATP-dependent NAD(P)HX dehydratase; ATP-dependent NAD(P)HX dehydratase {ECO:0000255; carbohydrate kinase domain containing; Carbohydrate kinase domain-containing protein; carbohydrate kinase domain-containing protein {ECO:0000255; CARKD; HAMAP-Rule:MF_03157}; LP3298; NAD(P)HX dehydratase; Naxd; RGD1562691; zgc:171429
Gene Symbols NAXD
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CARKD. Recombinant protein control fragment (Product #RP-89847).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55739
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.