Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CARKD Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543038
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543038 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543038 Supplier Invitrogen™ Supplier No. PA543038
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: RLHALVVGPG LGRDDALLRN VQGILEVSKA RDIPVVIDAD GLWLVAQQPA Sequence homology: Cow: 100%; Dog: 92%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 92%; Rat: 100%; Yeast: 85%; Zebrafish: 77%.

Catalyzes the dehydration of the S-form of NAD(P)HX at the expense of ATP, which is converted to ADP. Together with NAD(P)HX epimerase, which catalyzes the epimerization of the S- and R-forms, the enzyme allows the repair of both epimers of NAD(P)HX, a damaged form of NAD(P)H that is a result of enzymatic or heat-dependent hydration. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CARKD
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene NAXD
Gene Accession No. Q8IW45
Gene Alias 0710008K08Rik; 2810407E01Rik; ATP-dependent (S)-NAD(P)H-hydrate dehydratase; ATP-dependent (S)-NAD(P)H-hydrate dehydratase {ECO:0000255; ATP-dependent NAD(P)H-hydrate dehydratase; ATP-dependent NAD(P)HX dehydratase; ATP-dependent NAD(P)HX dehydratase {ECO:0000255; carbohydrate kinase domain containing; Carbohydrate kinase domain-containing protein; carbohydrate kinase domain-containing protein {ECO:0000255; CARKD; HAMAP-Rule:MF_03157}; LP3298; NAD(P)HX dehydratase; Naxd; RGD1562691; zgc:171429
Gene Symbols NAXD
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human CARKD.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55739
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.