Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carmine, calcium/aluminum lake, Thermo Scientific™

Catalog No. AAA1665906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1665906 5 g
AAA1665914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1665906 Supplier Thermo Scientific Chemicals Supplier No. A1665906

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1390-65-4
Molecular Formula C22H20O13
Molecular Weight (g/mol) 492.39
MDL Number MFCD00167028
InChI Key DGQLVPJVXFOQEV-UHFFFAOYNA-N
Synonym carmine, b rose liquid, cochineal extract lake, carmine coccus cacti l., cochineal coccus cacti l., fema no. 2242, fema no. 2330, ccris 1204, alum lake
PubChem CID 25113282
IUPAC Name 3,5,6,8-tetrahydroxy-1-methyl-9,10-dioxo-7-[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]-9,10-dihydroanthracene-2-carboxylic acid
SMILES CC1=C(C(O)=O)C(O)=CC2=C1C(=O)C1=C(O)C(C3OC(CO)C(O)C(O)C3O)=C(O)C(O)=C1C2=O

Specifications

Quantity 5 g
Merck Index 14,1843
Chemical Name or Material Carmine, calcium/aluminum lake
RTECSNumber FH8891000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.