Learn More
StatLab Carnoy’s 2000 (Chloroform Free), 1 Gallon
SDP
Supplier:  StatLab FXCAR2GAL
Encompass_Preferred
Carnoy’s 2000™ solution fixes glycogen, Nissl substance, plasma cells, RNA, and DNA without Chloroform, Methanol, or Mercuric Chloride.
Catalog No. NC0207160
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.