Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Elodea densa
SDPSCI_ED

Catalog No. S07498ND Shop All Carolina Biological Supply Products
Change view
Click to view available options
Product Type:
Elodea densa
Elodea densa Coupon
Quantity:
12/Container
25/Container
50/Container
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Quantity
S07498ND Elodea densa 25/Container
S07497ND Elodea densa 12/Container
S07496ND Elodea densa Coupon 12/Container
S07499ND Elodea densa 50/Container
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Catalog No. S07498ND Supplier Carolina Biological Supply Supplier No. 162102

This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-766-7000. Provide the NEED BY DATE to the customer service agent.

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

Your fish can breathe easier with Elodea densa, the best of the oxygenating plants.

This vigorous, healthy plant needs good light and should be anchored in soil or sand.

  • Cultivated form
  • Keeps water conditioned and oxygenated
  • Can survive and grow as floating fragments
  • Recommended for photosynthesis studies
  • With instructions

Note: Prepaid coupons allow you to order your organisms in advance. We'll send a coupon(s) that you later redeem by phone, fax, Online web form, or mail. To ensure prompt arrival of material, coupons must be redeemed at least 2 weeks prior to requested delivery date.

Specifications

For Use With (Application) Photosynthesis studies
Quantity 25/Container
Scientific Name Elodea densa
Includes Instructions
Sufficient For Class of 25 students
Product Type Elodea densa
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.