Learn More
Description
Fungi are important decomposers, helping recycle organic materials in food webs, which makes them great subjects for ecological studies. Students can culture them for genetic studies. For studies in biochemistry, students can investigate the role yeast plays in the production of wine, beer and baked goods.
- Available in tube cultures, plate cultures or MicroKwik Cultures™
- Labeled by name and media on which they are cultivated
- Guaranteed for purity and genera
- Incubate at room temperature (26°C)
Carolina™ MicroKwick Cultlure™: Since a MicroKwik Culture can be refrigerated, you can order it ahead of time for use on short notice.
Specifications
Specifications
| For Use With (Application) | Teach many topics in ecology, genetics, biochemistry and other disciplines. |
| Packaging | Tube |
| Storage Requirements | 26°C |
| Quantity | Ea. |
| Scientific Name | Mucor hiemalis+ |
| Includes | Carolina™ techniques for studying bacteria and fungi manual |
| Sufficient For | Class of 30 students |
| Media Type | Potato Dextrose Agar |
| Product Type | Bacteria Culture |
| Temperature (Metric) Range | 25°C |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

