Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Individual Fungi Cultures
SDPSCI_ED

Catalog No. S07437ND Shop All Carolina Biological Supply Products
Change view
Click to view available options
Media Type:
Potato Dextrose Agar
Rabbit Dung Agar
Packaging:
Plate
Tube
Vial
Scientific Name:
Aspergillus niger
Mucor hiemalis+
Penicillium notatum
Pilobolus crystallinus
Rhizopus stolonifer (+)
Storage Requirements:
20°C to 22°C
26°C
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Media Type Packaging Scientific Name Storage Requirements
S07437ND Rabbit Dung Agar Plate Pilobolus crystallinus 26°C
S07241ND Potato Dextrose Agar Tube Mucor hiemalis+ 26°C
S07207ND Potato Dextrose Agar Tube Rhizopus stolonifer (+) 20°C to 22°C
S07438ND Potato Dextrose Agar Plate Rhizopus stolonifer (+) 26°C
S07206ND Potato Dextrose Agar Tube Penicillium notatum 20°C to 22°C
S07205ND Potato Dextrose Agar Tube Aspergillus niger 20°C to 22°C
S07243ND Potato Dextrose Agar Vial Penicillium notatum 26°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Catalog No. S07437ND Supplier Carolina Biological Supply Supplier No. 156207

This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-766-7000. Provide the NEED BY DATE to the customer service agent.

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

Fungi can help you teach many topics in ecology, genetics, biochemistry and other disciplines.

Fungi are important decomposers, helping recycle organic materials in food webs, which makes them great subjects for ecological studies. Students can culture them for genetic studies. For studies in biochemistry, students can investigate the role yeast plays in the production of wine, beer and baked goods.

  • Available in tube cultures, plate cultures or MicroKwik Cultures™
  • Labeled by name and media on which they are cultivated
  • Guaranteed for purity and genera
  • Incubate at room temperature (26°C)

Carolina™ MicroKwick Cultlure™: Since a MicroKwik Culture can be refrigerated, you can order it ahead of time for use on short notice.

Specifications

For Use With (Application) Teach many topics in ecology, genetics, biochemistry and other disciplines.
Packaging Plate
Storage Requirements 26°C
Quantity Ea.
Scientific Name Pilobolus crystallinus
Includes Carolina™ techniques for studying bacteria and fungi manual
Sufficient For Class of 30 students
Media Type Rabbit Dung Agar
Product Type Bacteria Culture
Temperature (Metric) Range 25°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.