Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Nutrient Agar, Prepared Media Bottles
SDPSCI_ED

Catalog No. S21805ND Shop All Carolina Biological Supply Products
Change view
Click to view available options
Quantity:
125 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S21805ND 125 mL
S21808ND 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Catalog No. S21805ND Supplier Carolina Biological Supply Supplier No. 776360
Add to Cart
Add to Cart
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

Prepared Media Bottles - 100% Satisfaction Guaranteed!

The bottle is sealed to prevent contamination and drying. All bottles are glass, except where noted.

  • No autoclave needed
  • Free booklet on bottled media included
  • Note: C of A unavailable for this item

Specifications

Form Agar
Format Bottle
Manufacturing Origin United States
Media Type Nutrient Agar
Product Type Prepared Microbiology Media
Sterility Sterile
Target Organism Class Fastidious and Non-fastidious Organisms
Test Type Microbial Cultivation
Formulation Each glass bottle of bacteriological medium contains 125mL of ready-to-use material, except where noted. This amount is sufficient for pouring up to 5 plates or 15 to 20 tubes.
Quantity 125 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.