Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Wild-Type Drosophila
SDPSCI_ED

Catalog No. S07675ND Shop All Carolina Biological Supply Products
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
S07675ND 1 Ea.
1 options
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

Catalog No. S07675ND

Supplier: Carolina Biological Supply 172100

This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-766-7000. Provide the NEED BY DATE to the customer service agent.

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

The wild-type, Oregon R strain of Drosophila is the standard red eye fruit fly.

This culture is suitable as a parental strain, used along with one of the mutant strains to construct a heterozygous F1 generation and a subsequent F2 generation for study.

  • Short generation times
  • Easy to raise
  • Affordable
  • Minimal storage space needed
  • Sequenced genome
  • Easy for beginners to score
  • Appropriate for animal behavior studies

Specifications

For Use With (Application) Suitable as a parental strain, used along with one of the mutant strains to construct a heterozygous F1 generation and a subsequent F2 generation for study, these wild type flies are also appropriate for animal behavior studies.
Storage Requirements 20°C to 25°C
Quantity 1 Ea.
Sufficient For Class of 30 students
Product Type Drosophila, Wild Type
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.