Learn More
Carolina™ Carosafe™ Holding Solution



For use in formalin-preserved specimens. Carolina™ Carosafe™ Holding Solution is formaldehyde-free and cannot be used to fix specimens.
$52.50 - $178.50
Specifications
Product Type | Ready to use. |
---|---|
For Use With (Application) | Preservative and holding solution for biological specimens |
Alert:
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Description
- Carosafe preservative and holding solution for biological specimens contains no formaldehyde and is not a tissue fixative
- This produces a formalin-preserved specimen that, when dissected, minimizes formaldehyde exposure
Order Info
For use in science education laboratories – no certificate of analysis is available; Ready-to-use; do not dilute; Health Risk: 1; Flammability: 1; Reactivity: 0
Specifications
Ready to use. | |
Preservative and holding solution for biological specimens |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.