Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

β-Carotene 99%, Thermo Scientific™

Catalog No. AC404150010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC404150010 1 g
AC404150050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC404150010 Supplier Thermo Scientific Chemicals Supplier No. 404150010

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7235-40-7
Molecular Formula C40H56
Molecular Weight (g/mol) 536.89
MDL Number MFCD00001556
InChI Key OENHQHLEOONYIE-JLTXGRSLSA-N
Synonym beta-carotene, beta carotene, betacarotene, beta,beta-carotene, provitamin a, solatene, carotaben, provatene, all-trans-beta-carotene, serlabo
PubChem CID 5280489
ChEBI CHEBI:17579
IUPAC Name 1,3,3-trimethyl-2-[(1E,3E,5E,7E,9E,11E,13E,15E,17E)-3,7,12,16-tetramethyl-18-(2,6,6-trimethylcyclohex-1-en-1-yl)octadeca-1,3,5,7,9,11,13,15,17-nonaen-1-yl]cyclohex-1-ene
SMILES C\C(\C=C\C=C(/C)\C=C\C1=C(C)CCCC1(C)C)=C/C=C/C=C(\C)/C=C/C=C(\C)/C=C/C1=C(C)CCCC1(C)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Quantity 1 g
Merck Index 15, 1855
Formula Weight 536.88
Chemical Name or Material β-Carotene, 99%
Hazard Category LIGHT SENSITIVE
Hazard Statement MOISTURE SENSITIVE
Precautionary Statement AIR SENSITIVE
EINECSNumber 230-636-6
RTECSNumber FI0329500
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.