Learn More
Apexbio Technology LLC NAD+ 53-84-9 10mM (in 1mL H2O)
Supplier: Apexbio Technology LLC B17935.1
Nicotinamide adenine dinucleotide (NAD CAS number 53-84-9) is a coenzyme involved in oxidation-reduction reactions accepting electrons to form NADH Additionally NAD functions as a substrate in enzymatic processes mediated by poly(ADP-ribose) polymerases sirtuins and cyclic ADP-ribose synthases NAD analogs demonstrate inhibitory activity against CD38-associated NAD glycohydrolase (NADase) with reported IC50 values ranging from 2 mM to 10 mM In biomedical research NAD supplementation is investigated for addressing fatigue and fatigue-associated disorders
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.