Learn More
Research Products International Corp Carrageenan, Iota Type, 100 Grams
SDP
Supplier:  Research Products International Corp C41090100.0
Encompass_Preferred
Iota carrageenan binds water to form dry, elastic gels in the presence of calcium salts. Ca2+ establish bonds between the carrageenan molecules to form helices. The negative charges associated with the 2-sulfate groups on the iota carrageenan molecules do not allow the helices to aggregate to the same extent as Kappa carrageenan.
Catalog No. 50-213-245
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.