Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carrageenan, iota type

Catalog No. AAJ6060322
Change view
Click to view available options
Quantity:
100 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6060322 100 g
AAJ6060330 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6060322 Supplier Thermo Scientific Chemicals Supplier No. J6060322
Add to Cart

Forms soft gels in the presence of calcium ions | CAS: 9062-07-1 | (C12H16O15S2)n | NaN g/mol

Carrageenan, iota type is used as a stabilizer, for suspending cocoa in chocolate manufacture, and to clarify beverages. Carrageenans are used commercially as thickeners and stabilizing agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Carrageenan, iota type is used as a stabilizer, for suspending cocoa in chocolate manufacture, and to clarify beverages. Carrageenans are used commercially as thickeners and stabilizing agents.

Solubility
Partially soluble in water, Soluble in hot water. Insoluble in ethanol, in hot concentrated salt solutions.

Notes
Stable under normal conditions. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away fom strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9062-07-1
Molecular Formula (C12H16O15S2)n
Molecular Weight (g/mol) NaN
MDL Number MFCD00151512
InChI Key QIDSWKFAPCTSKL-RRQHLKGPSA-J
Synonym iota-carrageenans, 1->4-3,6-anhydro-2-o-sulfonato-alpha-d-galactopyranosyl-1->3-4-o-sulfonato-beta-d-galactopyranan
PubChem CID 11966245
IUPAC Name [(2R,3R,4R,5R,6S)-4,5-dihydroxy-6-[[(1R,3R,4R,5S,8S)-3-[(2R,3S,4R,5R,6S)-5-hydroxy-2-(hydroxymethyl)-6-[[(1R,3S,4R,5S,8S)-3-hydroxy-4-sulfonatooxy-2,6-dioxabicyclo[3.2.1]octan-8-yl]oxy]-3-sulfonatooxyoxan-4-yl]oxy-4-sulfonatooxy-2,6-dioxabicyclo[3.2.1]oct
SMILES OC[C@H]1O[C@@H](O[C@H]2[C@H]3CO[C@@H]2[C@@H](OS(O)(=O)=O)[C@@H](-*)O3)[C@H](O)[C@@H](O-*)[C@H]1OS(O)(=O)=O

Specifications

Color Brown to White
Quantity 100 g
Solubility Information Partially soluble in water,Soluble in hot water. Insoluble in ethanol,in hot concentrated salt solutions.
Physical Form Powder
Chemical Name or Material Carrageenan, iota type
EINECSNumber 232-949-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.