Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CARS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559722
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559722 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559722 Supplier Invitrogen™ Supplier No. PA559722
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VFGAIISYFE QFFETVGISL ANQQYVSGDG SEATLHGVVD ELVRFRQKVR QFALAMPEAT GDARRQQLLE RQPLLEACDT LRRGLTAHGI NIKDRSSTTS TWELLDQR Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 73%.

CARS is a class 2 aminoacyl-tRNA synthetase, cysteinyl-tRNA synthetase. Each of the twenty aminoacyl-tRNA synthetases catalyzes the aminoacylation of a specific tRNA or tRNA isoaccepting family with the cognate amino acid. It catalyzes the chemical reaction: ATP + L-cysteine + tRNA(Cys) = AMP + diphosphate + L-cysteinyl-tRNA(Cys).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CARS2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CARS2
Gene Accession No. Q9HA77
Gene Alias CARS2; COXPD27; cysRS; cysteine tRNA ligase 2, mitochondrial (putative); Cysteinyl-tRNA synthetase; cysteinyl-tRNA synthetase 2, mitochondrial (putative); OK/SW-cl.10; Probable cysteine--tRNA ligase, mitochondrial
Gene Symbols CARS2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CARS2. Recombinant protein control fragment (Product #RP-97959).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79587
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.