Learn More
CPC Scientific H-Val-Pro-Ile-Tyr-Glu-Lys-Lys-Tyr-Gly-Gln-Val-Pro-Met-Cys-Asp-Ala-Gly-Glu-Gln-Cys-Ala-Val-Arg-Lys-Gly-Ala-Arg-Ile-Gly-Lys-Leu-Cys-Asp-Cys-Pro-Arg-Gly-Thr-Ser-Cys-Asn-Ser-Phe-Leu-Leu-Lys-Cys-Leu-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific CART004B
Encompass
SEQUENCE: H-Val-Pro-Ile-Tyr-Glu-Lys-Lys-Tyr-Gly-Gln-Val-Pro-Met-Cys-Asp-Ala-Gly-Glu-Gln-Cys-Ala-Val-Arg-Lys-Gly-Ala-Arg-Ile-Gly-Lys-Leu-Cys-Asp-Cys-Pro-Arg-Gly-Thr-Ser-Cys-Asn-Ser-Phe-Leu-Leu-Lys-Cys-Leu-OH (trifluoroacetate salt)(Cys20 and 40, Cys14 and 32, Cys34 and 47 bridge)ONE-LETTER SEQUENCE: VPIYEKKYGQVPMCDAGEQCAVRKGARIGKLCDCPRGTSCNSFLLKCL (Cys20 and 40 bridge, Cys14 and 32 bridge, Cys34 and 47 bridge)MOLECULAR FORMULA: C225H365N65O65S7MOLECULAR WEIGHT: 5245.23STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [214050-22-3]SYNONYMS: CART (55-102) (human)RESEARCH AREA: ObesityREFERENCES:Murphy KG. Brief Funct Genomic Proteomic. 2005, 4(2):95-111.SKU(s): CART-004A, CART-004B
Catalog No. 50-210-8924
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.