Learn More
ORS Nasco CART MERC VAP HEPA P100
Supplier:  ORS Nasco 06875852P100L
Encompass_Preferred
Honeywell part# 75852P100L. Honeywell North Low-Profile Cartridges and Cartridge/P100 Filter Combinations Cartridge Type: Cartridge/Filter Assembly Olive Magenta Quantity: 12 Case Applications: Industrial applications For Use With: Respirator Mercury Vapor Chlorine And P100 Certifications/Compliance: NIOSH 42CFR84
Catalog No. 50-255-1668
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.