Learn More
CPC Scientific H-Ile-Pro-Ile-Tyr-Glu-Lys-Lys-Tyr-Gly-Gln-Val-Pro-Met-Cys-Asp-Ala-Gly-Glu-Gln-Cys-Ala-Val-OH (trifluoroacetate salt) 1MG
Supplier: CPC Scientific CART001B
ONE-LETTER SEQUENCE: IPIYEKKYGQVPMCDAGEQCAV (Cys14 and 20 bridge)MOLECULAR FORMULA: C107H166N26O33S3MOLECULAR WEIGHT:2440.82STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [1815618-15-5], SYNONYMS: CART (55-76) (rat)RESEARCH AREA: Obesity
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.