Learn More
Cayman Chemical CarteololhydrochlorIde 250mg
SDP
Supplier:  Cayman Chemical 23758250 MG
Encompass_Preferred
An inhibitor of the Bcl-2 family proteins Bcl-2 Mcl-1 and Bcl-xL (Kis 120 260 and 1100 nM respectively) induces apoptosis inhibits migration and capillary sprouting in endothelial cells has apoptotic action against leukemia lymphoma and pancreatic cancer cells
Catalog No. 50-299-4557
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.