Learn More
Thermo Scientific CARTRIDGE 13.5ML
Supplier:  Thermo Scientific 79252
Encompass_Preferred
Rotors for Centrifuges; Quick-Seal individual cartridge; Thermo Scientific; for Stepsaver 65V13 vertical titanium rotor; 13.5mL; Each
Catalog No. 50-867-257