Learn More
Thermo Scientific CARTRIDGE13.5/39ML UC(1PER)
Supplier:  Thermo Scientific 79263
Encompass_Preferred
Centrifuge Accessories; Cartridges; Thermo Scientific; For use with 50V39 vertical titanium rotor; Accommodates 16 x 93mm, 13.5mL Ultracrimp tubes; Individual
Catalog No. 50-867-262