Learn More
Thermo Scientific CARTRIDGE39MLUC8PER
Supplier:  Thermo Scientific 79299
Encompass_Preferred
Centrifuge Accessories; Cartridges; Thermo Scientific; For use with 50V39 vertical titanium rotor; Accommodates 25 x 113mm, 39mL Ultracrimp tubes; Set of 8
Catalog No. 50-867-273