Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3M™ 6000 Series Cartridges
NIOSH-approved cartridges for use with 3M™ full and half facepiece respirators 6000, 7000 and FF-400 Series.
Supplier: 3M™ 7000001937
This item is not returnable.
View return policy
Description
NIOSH-approved cartridges for use with 3M™ full and half facepiece respirators 6000, 7000 and FF-400 Series.
- For use with 3M full and half facepiece respirators 6000, 7000 and FF-400 Series
- Bayonet cartridge connection
- NIOSH-approved for protection against a variety of gases, vapors, and particulate hazards*
- For contaminants up to 10X the Permissible Exposure Limit (PEL) with half-facepiece respirators or 50X the PEL (when fit tested) with full-facepiece respirators or according to specific OSHA standards or applicable government regulations, whichever is lower
- Not intended for use in atmospheres that are unknown or immediately dangerous to life or death (IDLH)
- Do not exceed maximum use concentrations established by regulatory standards
- These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product
- Misuse may result in sickness or death
*Details found at individual product level according
Specifications
| Ammonia, Methylamine | |
| Bayonet | |
| Cartridge | |
| 3M™ half and full facepiece 6000, 7000, ff-400 series respirators | |
| 60/Cs. | |
| 10.6 cm | |
| 8.6 cm |
| NIOSH-approved | |
| Green | |
| Demolition, facility maintenance, sanding | |
| 6004 | |
| 4.2 in. | |
| 3.4 in. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction