Learn More
Thermo Scientific™ Barnstead™ Water System Cartridges
Catalog No. 10451027
Change view
Click to view available options
Description:
High purity water cartridge
Ultrapure polisher cartridge
For Use With (Equipment):
GenPure GenPure Pro, GenPure xCAD, LabTower EDI
MicroPure
Pacific TII
Smart2Pure
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description For Use With (Equipment)
10-451-027 Ultrapure polisher cartridge MicroPure
10-451-042 High purity water cartridge Pacific TII
10-451-028 Ultrapure polisher cartridge Smart2Pure
10-451-033 Ultrapure polisher cartridge GenPure GenPure Pro, GenPure xCAD, LabTower EDI
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 10-451-027 Supplier Thermo Scientific™ Supplier No. 09.1006
Add to Cart
Edge
Add to Cart
Maintain your Thermo Scientific™ Barnstead™ point of use water purification system with these replacement cartridges.
Maintain your Thermo Scientific™ Barnstead™ point of use water purification systems with replacement Thermo Scientific™ Barnstead™ Cartridges.
Choose the appropriate part number for your system. These cartridges are for use with the following Barnstead point of use water purification systems:
  • MicroPure™
  • GenPure™
  • GenPure™ Pro
  • GenPure™ xCAD
  • Smart2Pure™
  • LabTower™ EDI
  • Pacific™ TII
TRUSTED_SUSTAINABILITY
Specifications
Description Ultrapure polisher cartridge
For Use With (Equipment) MicroPure
Type Cartridge, Ultrapure Polisher