Learn More
Enzo Life Sciences Carvedilol (100mg). CAS: 72956-09-3
SDP
Supplier:  Enzo Life Sciences BMLAR1120100
Encompass_Preferred
An antagonist at α1- and β-adrenergic receptors. Displays antioxidant and antiapoptotic effects in vivo. Alternative name: 1-(9H-Carbazol-4-yloxy)-3- 2-(2-methoxyphenoxy)ethylamino propan-2-ol. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (>25mg/ml) or 100% ethanol (10mg/ml). Long Term Storage: +4°C.
Catalog No. 50-201-0327
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.