Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carvedilol 98.0+%, TCI America™
SDP

Catalog No. C226025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C226025G 25 g
C22605G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C226025G Supplier TCI America Supplier No. C226025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 72956-09-3
Molecular Formula C24H26N2O4
Molecular Weight (g/mol) 406.48
MDL Number MFCD00864692,MFCD00869663
InChI Key OGHNVEJMJSYVRP-UHFFFAOYNA-N
Synonym carvedilol, coreg, dilatrend, carvedilolum, eucardic, carvedilolum latin, kredex, querto, 1-9h-carbazol-4-yloxy-3-2-2-methoxyphenoxy ethyl amino-2-propanol, 1-9h-carbazol-4-yl oxy-3-2-2-methoxyphenoxy ethyl amino propan-2-ol
PubChem CID 2585
ChEBI CHEBI:3441
IUPAC Name 1-(9H-carbazol-4-yloxy)-3-{[2-(2-methoxyphenoxy)ethyl]amino}propan-2-ol
SMILES COC1=CC=CC=C1OCCNCC(O)COC1=CC=CC2=C1C1=C(N2)C=CC=C1

Specifications

Melting Point 117°C
Color White-Yellow
Quantity 25 g
UN Number 3077
Formula Weight 406.48
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Carvedilol
RTECSNumber UA8670000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.