Learn More
Thermo Scientific™ Cary-Blair with Indicator Single Vial
Description
Collect, preserve, and recover enteric bacterial pathogens in stool specimens with Thermo Scientfic™ Cary-Blair with Indicator Single Vial, part of a complete collection of quality fecal transports which come with industry standard color-coding and easy-to-use sporked caps.
- Cary-Blair with Indicator is an isotonic and nonnutritive medium, which does not contain a fermentable carbohydrate.
- Sodium thioglycollate is added to impede oxidation.
- Disodium phosphate is incorporated in the medium to act as a buffering agent, which prevents overgrowth of Escherichia coli and Enterobacter species.
- The relatively high pH minimizes the destruction of bacteria due to acid formation.
- Phenol red, a pH indicator, is added to demonstrate acidic shifts that can affect the viability of some enteric pathogens.
Add convenience and safety to your fecal collection procedures with a broad line of transport products which have a long shelf life and are designed especially for enteric diagnostic testing.
Look to us for convenient kit packaging options, including bulk kits.
Multilingual patient collection instructions are included.
Choose from a wide range of collection and preservation products for fecal specimens, which is important to the successful growth, isolation and detection of microorganisms.
- Cary Blair with Indicator
- Formalin 10%
- Formalin 5%
- PVA Modified Fixative
- SAF Fixative
- Zinc PVA Fixative
- Empty Vial
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Specifications
| Product Type | Fecal Transport Vial |
| Closure Color | Orange |
| Description | Cary-Blair with Indicator Single Vial (Orange Cap) |
| Fill Volume | 15 mL |
| Media Type | Cary-Blair |
| Packaging | Single vial. Packaged in partitioned boxes. |
| Quantity | 120 Vials/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.