Learn More
Invitrogen™ Cas12j Monoclonal Antibody (5F95)

Description
Cas12j Monoclonal Antibody for Western Blot, ICC/IF
Specifications
Specifications
| Antigen | Cas12j |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 5F95 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 5mM sodium azide |
| Gene Alias | Cas theta |
| Host Species | Mouse |
| Immunogen | Recombinant Cas theta/Cas-12j expressed in and purified from E. coli. |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.