Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CASC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113041
Change view
Click to view available options
Quantity:
100 μg
Catalog No. Quantity
PIPA5113041 100 μg
1 options

Catalog No. PIPA5113041

Supplier: Invitrogen™ PA5113041

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CASC1 Polyclonal Antibody for Western Blot, IHC (P), ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen CASC1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene CASC1
Gene Accession No. Q6TDU7
Gene Alias cancer susceptibility candidate 1; cancer susceptibility candidate gene 1 protein; CASC1; LAS1; lung adenoma susceptibility 1; lung adenoma susceptibility 1-like protein; PPP1R54; Protein CASC1; protein phosphatase 1 regulatory subunit 54; protein phosphatase 1, regulatory subunit 54
Gene Symbols CASC1
Host Species Rabbit
Immunogen Recombinant Human Protein CASC1 protein (1-300AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 297720, 55259
Target Species Human, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.