Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Casein Blocking Buffer, 3% in PBS

Catalog No. AAJ60289AP
Change view
Click to view available options
Quantity:
250 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60289AP 500 mL
AAJ60289AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60289AP Supplier Thermo Scientific Chemicals Supplier No. J60289AP
Add to Cart

missing translation for 'validMainframeMsg'

For Western blots and ELISA

The phosphate- and Tris-buffered saline formulations are used for blocking excess binding sites on membranes and microplates in antibody-based detection methods in ELISA, Western blotting, immunohistochemistry and other immunochemical applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Blocking Buffer
Concentration 3% in PBS
For Use With (Application) Western Blot
Physical Form Viscous Liquid
Quantity 500 mL

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.