Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Casein Kinase 1 gamma Antibody [PerCP], R&D Systems™
SDP

Catalog No. AF3186C
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3186C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AF3186C Supplier R&D Systems Supplier No. AF3186C

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Sheep Polyclonal Antibody

Casein Kinase 1 gamma Polyclonal specifically detects Casein Kinase 1 gamma in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blot.

Specifications

Antigen Casein Kinase 1 gamma
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate PerCP
Formulation PBS
Gene Alias casein kinase 1, gamma 1, casein kinase I isoform gamma-1, CKI-gamma 1, EC 2.7.11, EC 2.7.11.1
Host Species Sheep
Immunogen E. coli-derived recombinant human CK1 gamma , Val182-Tyr304, Accession # Q9HCP0
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 53944
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.