Learn More
Medchemexpress LLC HY-123954 5mg , Casein Kinase inhibitor A51 CAS:2079068-74-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1239545MG
Encompass_Preferred
HY-123954 5mg Casein Kinase inhibitor A51 CAS: 2079068-74-7 Casein Kinase inhibitor A51 is a potent and orally active casein kinase 1α (CK1α) inhibitor. Casein Kinase inhibitor A51 induces leukemia cell apoptosis, and has potent anti-leukemic activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9659
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.