Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Casein, MP Biomedicals™

Catalog No. MP219990901 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 lb.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219990901 1 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP219990901 Supplier MP Biomedicals™ Supplier No. 0219990901
Add to Cart
Add to Cart

Casein is Vitamin-tested and micropulverized. Product composed of thiamin, riboflavin, niacin, pyridoxine, folic acid, Vitamin B12, biotin, and pantothenic acid.

TRUSTED_SUSTAINABILITY

Specifications

Grade Vitamin Tested, Micropulverized
Content And Storage Store at Room Temperature (15–30°C).
Quantity 1 lb.
Molecular Weight (g/mol) 75,000-100,000g/mol
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.