Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Casein Peptone

Catalog No. 113066132 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity: 1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
11-306-6132 Quantity: 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-306-6132 Supplier MP Biomedicals™ Supplier No. 113066132
Add to Cart
Add to Cart

Casein Peptone is supplemented to microbiological growth media.

  • Casein Peptone, Type I
  • Molecular Biology Certified™
TRUSTED_SUSTAINABILITY

Specifications

Form Powder
Grade Molecular Biology Certified™
For Use With (Application) Cell Culture, Proteomics, Purification and Detection, Recombinant Protein Expression and Analysis, Bacterial Culture, Plasmid Production, Protein Expression, Gene Expression, Cloning, Genetic Manipulation, Growth and Maintenance of M13 Phage and Other Filamentous ssDNA Bacteriophages
pH 6.5 to 7.5 (2% aq. soln.)
Quantity 1 kg
Solubility Soluble in water.
Synonym Amino Acid Hydrolysate Powder

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.