Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CASK Polyclonal Antibody, Huabio

Catalog No. 502544707
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4707 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4707 Supplier HUABIO RESEARCH INC Supplier No. ER180362
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CASK Polyclonal antibody specifically detects CASK in Human, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen CASK
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene Cask
Gene Accession No. O14936, Q62915
Gene Alias CAGH39; calcium/calmodulin dependent serine protein kinase; calcium/calmodulin-dependent serin protein kinase; Calcium/calmodulin-dependent serine protein kinase; calcium/calmodulin-dependent serine protein kinase (MAGUK family); calcium/calmodulin-dependent serine protein kinase membrane-associated guanylate kinase; CAMGUK; CASK; CMG; DXPri1; DXRib1; FGS4; G-protein-coupled receptor GPR34; hCASK; LIN2; LIN-2; MGC7449; MICPCH; mLin-2; MRXSNA; OTTHUMP00000023157; OTTHUMP00000023161; OTTHUMP00000023162; Pals3; peripheral plasma membrane protein CASK; Protein lin-2 homolog; RP23-278L4.1; TNRC8; trinucleotide repeat containing 8
Host Species Rabbit
Immunogen Recombinant protein within human CASK aa 300-500.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29647, 8573
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.