Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Caspase 1 Monoclonal Antibody (5B10), eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 5013821
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-138-21 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-138-21 Supplier Invitrogen™ Supplier No. 14983282
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rat Monoclonal Antibody

Description: The monoclonal antibody 5B10 recognizes mouse Caspase-1, a member of the cysteine-requiring aspartate protease family, which plays a critical role in cell suicide known as apoptosis. In particular, it converts pro-IL-1beta to mature IL-1beta; hence it is also called ICE (interleukin-1beta converting enzyme). Upon activation Caspase-1 exists as a tetramer of two p20 and two p10 subunits. There are several isoforms of Caspase-1. The 5B10 antibody recognizes both the pro-form of about 42kDa and the active p20 form. Applications Reported: This 5B10 antibody has been reported for use in immunoblotting (WB). Applications Tested: This 5B10 antibody has been tested by western blot analysis of lysates prepared from the J774 cell line. This can be used at 1-5 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

Caspases are a family of cysteine proteases that can be divided into the apoptotic and inflammatory caspase subfamilies. Unlike the apoptotic caspases, members of the inflammatory subfamily are generally not involved in cell death but are associated with the immune response to microbial pathogens. Members of this subfamily include caspase-1, -4, -5, and -12 and can activate proinflammatory cytokines such as IL-1-b and IL-18. Caspase-1 was initially identified as an IL-1-b-converting enzyme; later experiments revealed it to be a mammalian homolog of the C. elegans cell death gene ced-3 whose overexpression can induce apoptosis in fibroblasts.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Caspase 1
Applications Western Blot
Classification Monoclonal
Clone 5B10
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene CASP1
Gene Accession No. P29452
Gene Alias CASP1; CASP-1; CASP1 nirs variant 1; caspase 1; caspase 1, apoptosis-related cysteine peptidase (interleukin 1, beta, convertase); Caspase1; caspase-1; Caspase-1 subunit p10; Caspase-1 subunit p20; ICE; IL-1 beta-converting enzyme; IL-1B converting enzyme; IL1BC; IL-1BC; IL1BCE; IL1B-convertase; interleukin 1 beta-converting enzyme; interleukin 1, beta, convertase; interleukin 1-B converting enzyme; Interleukin 1beta converting enzyme; interleukin-1 beta convertase; Interleukin-1 beta-converting enzyme; p45
Gene Symbols CASP1
Host Species Rat
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12362
Target Species Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.