Learn More
Pro Sci Inc Caspase-12 Antibody (Large), Rabbit Polyclonal
SDP
Supplier:  Pro Sci Inc 3195
Encompass_Preferred
Caspase-12 Antibody (Large), Rabbit Polyclonal Validated in E, WB, IHC-P, IF and working on Human, Mouse, Rat
Catalog No. NC1857091
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.